Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,7-Methylenedioxy-4-methyl-3-maleimidocoumarin 98.0+%, TCI America™
SDP

Catalog No. M0881100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0881100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0881100MG Supplier TCI America Supplier No. M0881100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97744-90-6
Molecular Formula C15H9NO6
Molecular Weight (g/mol) 299.238
MDL Number MFCD00059921
InChI Key NSFINZUKLACTRX-UHFFFAOYSA-N
Synonym 3-Maleimido-4-methyl-6,7-methylenedioxycoumarin
PubChem CID 44630383
IUPAC Name 1-(8-methyl-6-oxo-[1,3]dioxolo[4,5-g]chromen-7-yl)pyrrole-2,5-dione
SMILES CC1=C(C(=O)OC2=CC3=C(C=C12)OCO3)N4C(=O)C=CC4=O

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 299.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Methylenedioxy-4-methyl-3-maleimidocoumarin [for HPLC Labeling]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.