accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3ONOH

CAS RN 97928-18-2

IUPAC Name: 1-(4-tert-butylphenyl)-4-(4-hydroxypiperidin-1-yl)butan-1-one
Synonyms: 1-(4-tert-butylphenyl)-4-(4-hydroxypiperidin-1-yl)butan-1-one
Molecular Weight (g/mol): 303.45
Molecular Formula: C19H29NO2
InChi Key: HUFTWIJFTZKVMR-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(=O)CCCN1CCC(O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results