accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CSOOClCH3SOOClCH3

CAS RN 97997-76-7

IUPAC Name: tetramethylbenzene-1,3-disulfonyl dichloride
Synonyms: tetramethylbenzene-1,3-disulfonyl dichloride
Molecular Weight (g/mol): 331.22
Molecular Formula: C10H12Cl2O4S2
InChi Key: YEYQIYAYJJXKFB-UHFFFAOYSA-N
SMILES: CC1=C(C)C(=C(C)C(=C1C)S(Cl)(=O)=O)S(Cl)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results