Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5,6-Tetramethylbenzenedisulfonyl Dichloride 98.0+%, TCI America™
SDP

Catalog No. T15625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T15625G Supplier TCI America Supplier No. T15625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97997-76-7
Molecular Formula C10H12Cl2O4S2
Molecular Weight (g/mol) 331.22
MDL Number MFCD00191502
InChI Key YEYQIYAYJJXKFB-UHFFFAOYSA-N
Synonym Isodurenedisulfonyl Dichloride

Specifications

Melting Point 136°C
Quantity 5 g
UN Number 3261
Formula Weight 331.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5,6-Tetramethylbenzenedisulfonyl Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.