accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98-11-3

IUPAC Name: benzenesulfonic acid
Synonyms: benzenesulfonic acid
Molecular Weight (g/mol): 158.17
Molecular Formula: C6H6O3S
InChi Key: SRSXLGNVWSONIS-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (9)
  • (6)
Quantity 
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Grade 
  • (10)
Percent Purity 
  • (1)
  • (5)
  • (3)
Physical Form 
  • (2)
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (6)
  • (1)
  • (13)
  • (5)
  • (9)
  • (6)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (10)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (5)