accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CH3CCH3

CAS RN 98-19-1

IUPAC Name: 1-tert-butyl-3,5-dimethylbenzene
Synonyms: 1-tert-butyl-3,5-dimethylbenzene
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: FZSPYHREEHYLCB-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)