Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-tert-Butyl-m-xylene, MP Biomedicals

Catalog No. ICN209611 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN209611 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN209611 Supplier MP Biomedicals, Inc Supplier No. 0520961180

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98-19-1
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
InChI Key FZSPYHREEHYLCB-UHFFFAOYSA-N
Synonym 5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl
PubChem CID 7378
IUPAC Name 1-tert-butyl-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)C(C)(C)C)C

Specifications

Quantity 100 g
Formula Weight 162.28
Chemical Name or Material 5-tert-Butyl-m-xylene
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.