accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98-30-6

IUPAC Name: 2-amino-4-methanesulfonylphenol
Synonyms: 2-amino-4-methanesulfonylphenol
Molecular Weight (g/mol): 187.21
Molecular Formula: C7H9NO3S
InChi Key: SFLMBHYNCSYPOO-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(O)C(N)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results