accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClSOHOO

CAS RN 98-36-2

IUPAC Name: 3-amino-4-chlorobenzene-1-sulfonic acid
Synonyms: 3-amino-4-chlorobenzenesulfonic acid
Molecular Weight (g/mol): 207.63
Molecular Formula: C6H6ClNO3S
InChi Key: XJQRCFRVWZHIPN-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1Cl)S(O)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results