accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CCH3

CAS RN 98-51-1

IUPAC Name: 1-tert-butyl-4-methylbenzene
Synonyms: P-(T-butyl)toluene
Molecular Weight (g/mol): 148.25
Molecular Formula: C11H16
InChi Key: QCWXDVFBZVHKLV-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)