Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butyltoluene, 95%

Catalog No. AAA18214AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA18214AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA18214AP Supplier Thermo Scientific Chemicals Supplier No. A18214AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 98-51-1 | C11H16 | 148.249 g/mol

4-tert-Butyltoluene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butyltoluene is used as pharmaceutical intermediate.

Solubility
Soluble in water (6 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-51-1
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00008837
InChI Key QCWXDVFBZVHKLV-UHFFFAOYSA-N
Synonym 4-tert-butyltoluene, p-tert-butyltoluene, 1-tert-butyl-4-methylbenzene, p-t-butyltoluene, benzene, 1-1,1-dimethylethyl-4-methyl, 4-t-butyltoluene, p-methyl-tert-butylbenzene, toluene, p-tert-butyl, 1-methyl-4-tert-butylbenzene, 4-methyl-tert-butylbenzene
PubChem CID 7390
IUPAC Name 1-tert-butyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)C(C)(C)C

Specifications

Melting Point -63°C
Density 0.853
Boiling Point 189°C to 193°C
Flash Point 62°C (144°F)
Odor Characteristic
Refractive Index 1.492
Quantity 500 mL
UN Number UN2667
Beilstein 2038670
Solubility Information Soluble in water (6g/L at 20°C).
Formula Weight 148.25
Percent Purity 95%
Chemical Name or Material 4-tert-Butyltoluene
Show More Show Less
Hazard Category H226-H302-H315-H319-H330-H335
Hazard Statement GHS H Statement
H330-H227-H302-H319-H335
Fatal if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 202-675-9
RTECSNumber XS8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.