accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOO

CAS RN 98-78-2

IUPAC Name: 3-oxocyclopentane-1-carboxylic acid
Synonyms: 3-oxocyclopentane-1-carboxylic acid
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: RDSNBKRWKBMPOP-UHFFFAOYNA-N
SMILES: OC(=O)C1CCC(=O)C1
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (4)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)