Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Oxocyclopentanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. O04161G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O04161G Supplier TCI America Supplier No. O04161G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98-78-2
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD01320173
InChI Key RDSNBKRWKBMPOP-UHFFFAOYNA-N
Synonym 3-oxocyclopentanecarboxylic acid, 3-oxo-cyclopentane-carboxylic acid, 3-oxo cyclopentane 1-carboxylic acid, 3-oxo-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-oxo, 3-carboxycyclopentanone, cyclopentanone-3-carboxylic acid, 3-oxo-cyclopentanecarboxylic acid, 3-ketocyclopentanecarboxylic acid, 3-cyclopentanone-1 r-carboxylic acid
PubChem CID 227798
ChEBI CHEBI:71213
IUPAC Name 3-oxocyclopentane-1-carboxylic acid
SMILES OC(=O)C1CCC(=O)C1

Specifications

Melting Point 62°C
Color White
Boiling Point 150°C
Quantity 1 g
Formula Weight 128.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Oxocyclopentanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.