accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98121-41-6

IUPAC Name: 5,6-dichloropyridin-3-amine
Synonyms: 5,6-dichloropyridin-3-amine
Molecular Weight (g/mol): 163
Molecular Formula: C5H4Cl2N2
InChi Key: XTCHZNJJTQACES-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(Cl)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results