Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2,3-dichloropyridine 98.0+%, TCI America™
SDP

Catalog No. A28245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28245G 5 g
A28241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28245G Supplier TCI America Supplier No. A28245G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 98121-41-6
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.00
MDL Number MFCD03840434
InChI Key XTCHZNJJTQACES-UHFFFAOYSA-N
PubChem CID 14595219
IUPAC Name 5,6-dichloropyridin-3-amine
SMILES NC1=CC(Cl)=C(Cl)N=C1

Specifications

Melting Point 113°C
Quantity 5 g
Formula Weight 163.00
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2,3-dichloropyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.