accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOO

CAS RN 98648-23-8

IUPAC Name: 4'-nitro-[1,1'-biphenyl]-4-carbaldehyde
Synonyms: 4'-nitro-[1,1'-biphenyl]-4-carbaldehyde
Molecular Weight (g/mol): 227.22
Molecular Formula: C13H9NO3
InChi Key: MGHIIGWDUVNPPV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(C=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results