accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99-12-7

IUPAC Name: 1,3-dimethyl-5-nitrobenzene
Synonyms: benzene, 1,3-dimethyl-5-nitro-
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: BYFNZOKBMZKTSC-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results