Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylnitrobenzene 98.0+%, TCI America™
SDP

Catalog No. D10865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D10865G Supplier TCI America Supplier No. D10865G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 99-12-7
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007269
InChI Key BYFNZOKBMZKTSC-UHFFFAOYSA-N
Synonym 5-nitro-m-xylene, 3,5-dimethylnitrobenzene, 3,5-dimethyl-1-nitrobenzene, benzene, 1,3-dimethyl-5-nitro, nitroxylol, m-xylene, 5-nitro, unii-5hvw2qtf0d, ccris 3121, 5hvw2qtf0d, 5-nitro-m-xylol
PubChem CID 7426
IUPAC Name 1,3-dimethyl-5-nitrobenzene
SMILES CC1=CC(=CC(C)=C1)[N+]([O-])=O

Specifications

Melting Point 73°C
Color Yellow
Boiling Point 273°C
Quantity 5 g
UN Number 3447
Formula Weight 151.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethylnitrobenzene
EINECSNumber (3)-0399
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.