accessibility menu, dialog, popup

UserName

Viewing profile as user:
OBiOOHOHHOO

CAS RN 99-26-3

IUPAC Name: 2,7-dihydroxy-2H-benzo[d]1,3-dioxa-2-bismacyclopentane-5-carboxylic acid
Synonyms: dermatol|bismuth subgallate|wismutgallathydroxid|gallic acid bismuth basic salt|basisches wismutgallat|basic bismuth gallate
Molecular Weight (g/mol): 394.09
Molecular Formula: C7H5BiO6
InChi Key: JAONZGLTYYUPCT-UHFFFAOYSA-K
SMILES: O[Bi]1OC2=C(O1)C(O)=CC(=C2)C(O)=O
Grade 
  • (3)
  • (9)
Percent Purity 
  • (3)
  • (3)
  • (5)
  • (1)
Molecular Weight (g/mol) 
  • (6)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (11)
  • (1)
  • (3)
  • (1)
  • (8)
  • (5)
  • (3)
  • (9)
  • (3)
  • (3)
  • (5)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)