Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subgallate (Powder/USP), Fisher Chemical

Catalog No. B338500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
B338-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B338-500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 99-26-3
Molecular Formula C7H5BiO6
Molecular Weight (g/mol) 394.09
MDL Number MFCD00044980
InChI Key JAONZGLTYYUPCT-UHFFFAOYSA-K
Synonym bismuth subgallate, wismutgallathydroxid, dermatol, bismuth gallate, dermatol puder, bismuth subgallas, bismutum subgallicum, basic bismuth gallate, gallic acid bismuth basic salt, wismutgallat, basisches
PubChem CID 16682999
IUPAC Name 2,7-dihydroxy-2H-benzo[d]1,3-dioxa-2-bismacyclopentane-5-carboxylic acid
SMILES O[Bi]1OC2=C(O1)C(O)=CC(=C2)C(O)=O

Specifications

Color Yellow
Density 1.10g/cm³
Loss on Drying 7.0% max.
Assay Percent Range 52.0 to 57.0 %
Packaging Poly Bottle
Quantity 500 g
Residual Solvents Meets Requirements
Formula Weight 394.09
Percent Purity 52.0 to 57.0%
Grade USP
Physical Form Powder or Solid
Chemical Name or Material Bismuth Subgallate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.