accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNOO

CAS RN 99-51-4

IUPAC Name: 1,2-dimethyl-4-nitrobenzene
Synonyms: 1,2-dimethyl-4-nitrobenzene
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: HFZKOYWDLDYELC-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1C)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)