Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethylnitrobenzene 98.0+%, TCI America™
SDP

Catalog No. D1085500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1085500G 500 g
D108525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1085500G Supplier TCI America Supplier No. D1085500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 99-51-4 | C8H9NO2 | 151.17 g/mol

Chemical Identifiers

CAS 99-51-4
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007268
InChI Key HFZKOYWDLDYELC-UHFFFAOYSA-N
Synonym 4-nitro-o-xylene, p-nitro-o-xylene, o-xylene, 4-nitro, benzene, 1,2-dimethyl-4-nitro, para-nitro-ortho-xylene, 3,4-dimethyl-1-nitrobenzene, 4-nitro-1,2-dimethylbenzene, 1,2-dimethyl-4-nitrobenzol, 3,4-dimethylnitrobenzene, ccris 3118
PubChem CID 7440
IUPAC Name 1,2-dimethyl-4-nitrobenzene
SMILES CC1=CC=C(C=C1C)[N+]([O-])=O

Specifications

Color Yellow
Quantity 500 g
Physical Form Crystal-Lump at 20°C
UN Number 3447
Formula Weight 151.17
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 3,4-Dimethylnitrobenzene
EINECSNumber (3)-0399
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.