CAS RN 99-53-6
IUPAC Name:
2-methyl-4-nitrophenol
Synonyms:
4-nitro-o-cresol
Molecular Weight (g/mol):
153.14
Molecular Formula:
C7H7NO3
InChi Key:
KDQPMQNHVQVVMR-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC=C1O)[N+]([O-])=O
1
–
2
of
2
results
1
4-Nitro-o-cresol 98.0+%, TCI America™
CAS: 99-53-6 Molecular Formula: C7H7NO3 Molecular Weight (g/mol): 153.14 MDL Number: MFCD01734013 InChI Key: KDQPMQNHVQVVMR-UHFFFAOYSA-N Synonym: 2-Hydroxy-5-nitrotoluene, 2-Methyl-4-nitrophenol PubChem CID: 7442 IUPAC Name: 2-methyl-4-nitrophenol SMILES: CC1=CC(=CC=C1O)[N+]([O-])=O
2
Sigma Aldrich 2-Methyl-4-nitrophenol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.