accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99-54-7

IUPAC Name: 1,2-dichloro-4-nitrobenzene
Synonyms: 1,2-dichloro-4-nitrobenzene
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: NTBYINQTYWZXLH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1
Boiling Point 
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)