Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloro-4-nitrobenzene, 99%, Thermo Scientific™

Catalog No. AAA1062030
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1062030 250 g
AAA106200B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1062030 Supplier Thermo Scientific Chemicals Supplier No. A1062030

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-54-7
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00007207
InChI Key NTBYINQTYWZXLH-UHFFFAOYSA-N
Synonym 3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech
PubChem CID 7443
IUPAC Name 1,2-dichloro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 39°C to 43°C
Boiling Point 255°C to 256°C
Flash Point 123°C (253°F)
Quantity 250 g
UN Number UN3077
Beilstein 1818163
Formula Weight 192
Percent Purity 99%
Chemical Name or Material 1,2-Dichloro-4-nitrobenzene
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-764-2
RTECSNumber CZ5250000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.