accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99-59-2

IUPAC Name: 2-methoxy-5-nitroaniline
Synonyms: 5-nitro-o-anisidine fast scarlet R azoamine scarlet K
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: NIPDVSLAMPAWTP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1N)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results