Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-nitroaniline, 99%, Thermo Scientific™

Catalog No. AC171305000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC171305000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC171305000 Supplier Thermo Scientific Chemicals Supplier No. 171305000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-59-2
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00007261
InChI Key NIPDVSLAMPAWTP-UHFFFAOYSA-N
Synonym 2-amino-4-nitroanisole, 5-nitro-o-anisidine, azoamine scarlet k, fast scarlet r, 2-methoxy-5-nitrobenzenamine, 2-amino-1-methoxy-4-nitrobenzene, azoamine scarlet, 5-nitro-2-methoxyaniline, o-anisidine nitrate, o-anisidine, 5-nitro
PubChem CID 7447
ChEBI CHEBI:48977
IUPAC Name 2-methoxy-5-nitroaniline
SMILES COC1=CC=C(C=C1N)[N+]([O-])=O

Specifications

Melting Point 116.0°C to 120.0°C
Linear Formula CH3OC6H3(NO2)NH2
Quantity 500 g
Solubility Information Solubility in water: soluble in hot water.
Formula Weight 168.15
Chemical Name or Material 2-Methoxy-5-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.