accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OH

CAS RN 99-71-8

IUPAC Name: 4-(butan-2-yl)phenol
Synonyms: 4-sec-butylphenol P-(sec-butyl)phenol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: ZUTYZAFDFLLILI-UHFFFAOYNA-N
SMILES: CCC(C)C1=CC=C(O)C=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results