Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-sec-Butylphenol 98.0+%, TCI America™
SDP

Catalog No. B073325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B073325G 25 g
B0733100G 100 g
B0733500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B073325G Supplier TCI America Supplier No. B073325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 99-71-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00002375
InChI Key ZUTYZAFDFLLILI-UHFFFAOYSA-N
PubChem CID 7453
ChEBI CHEBI:34442
IUPAC Name 4-butan-2-ylphenol
SMILES CCC(C)C1=CC=C(C=C1)O

Specifications

Melting Point 60°C
Color White
Boiling Point 110°C
Quantity 25 g
UN Number 2430
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-sec-Butylphenol
EINECSNumber (3)-0503
RTECSNumber SJ8924000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.