accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99-89-8

IUPAC Name: 4-(propan-2-yl)phenol
Synonyms: P-isopropylphenol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: YQUQWHNMBPIWGK-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(O)C=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (6)
  • (6)
Percent Purity 
  • (3)
  • (9)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (3)
  • (7)
  • (3)
  • (3)
  • (2)
  • (4)
  • (6)
  • (6)
  • (3)
  • (9)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)