accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99011-02-6

IUPAC Name: 1-(2-methylpropyl)-1H-imidazo[4,5-c]quinolin-4-amine
Synonyms: imiquimod imiquimod acetate aldara hydroxypropyl methylcellulose phthalate
Molecular Weight (g/mol): 240.31
Molecular Formula: C14H16N4
InChi Key: DOUYETYNHWVLEO-UHFFFAOYSA-N
SMILES: CC(C)CN1C=NC2=C1C1=CC=CC=C1N=C2N
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results