CAS RN 99076-56-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(p-Toluenesulfonyl)-L-alanine, 98%, Thermo Scientific™
CAS: 99076-56-9 MDL Number: MFCD02259513 Synonym: Tos-Ala-OH; N-Tosyl-L-alanine