accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3OOHNONHOOHO

CAS RN 99092-88-3

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-4-[(9H-xanthen-9-yl)carbamoyl]butanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-4-[(9H-xanthen-9-yl)carbamoyl]butanoic acid
Molecular Weight (g/mol): 426.47
Molecular Formula: C23H26N2O6
InChi Key: HOIIDGIJEPOSLL-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCC(=O)NC1C2=CC=CC=C2OC2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)