accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHOCH3OOCH3

CAS RN 992-42-7

IUPAC Name: [3-(hexadecanoyloxy)-5-(hydroxymethyl)-2-methylpyridin-4-yl]methyl hexadecanoate
Synonyms: [3-(hexadecanoyloxy)-5-(hydroxymethyl)-2-methylpyridin-4-yl]methyl hexadecanoate
Molecular Weight (g/mol): 646.01
Molecular Formula: C40H71NO5
InChi Key: PDJVTEPEYIYVJW-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OCC1=C(OC(=O)CCCCCCCCCCCCCCC)C(C)=NC=C1CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results