Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxine 3,4-Dipalmitate 98.0+%, TCI America™
SDP

Catalog No. P13955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13955G 5 g
P139525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13955G Supplier TCI America Supplier No. P13955G

Chemical Identifiers

CAS 992-42-7
Molecular Formula C40H71NO5
Molecular Weight (g/mol) 646.01
MDL Number MFCD01632152
InChI Key PDJVTEPEYIYVJW-UHFFFAOYSA-N
PubChem CID 10175530
IUPAC Name [3-hexadecanoyloxy-5-(hydroxymethyl)-2-methylpyridin-4-yl]methyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC1=C(C(=NC=C1CO)C)OC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 90°C
Color White
Quantity 5 g
Formula Weight 646.01
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridoxine 3,4-Dipalmitate
EINECSNumber (5)-3670
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.