accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaH3CNNNH2SOOONNNH2SOOOCH3(E/Z)(E/Z)

CAS RN 992-59-6

IUPAC Name: disodium 4-amino-3-(2-{4'-[2-(1-amino-4-sulfonatonaphthalen-2-yl)diazen-1-yl]-3,3'-dimethyl-[1,1'-biphenyl]-4-yl}diazen-1-yl)naphthalene-1-sulfonate
Synonyms: disodium 4-amino-3-(2-{4'-[2-(1-amino-4-sulfonatonaphthalen-2-yl)diazen-1-yl]-3,3'-dimethyl-[1,1'-biphenyl]-4-yl}diazen-1-yl)naphthalene-1-sulfonate
Molecular Weight (g/mol): 724.72
Molecular Formula: C34H26N6Na2O6S2
InChi Key: SUXCALIDMIIJCK-UHFFFAOYSA-L
SMILES: [Na+].[Na+].CC1=CC(=CC=C1N=NC1=CC(=C2C=CC=CC2=C1N)S([O-])(=O)=O)C1=CC=C(N=NC2=CC(=C3C=CC=CC3=C2N)S([O-])(=O)=O)C(C)=C1
Quantity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)