Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzopurpurin B, MP Biomedicals

Catalog No. ICN19506925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19506925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19506925 Supplier MP Biomedicals, Inc Supplier No. 0219506925

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 992-59-6
Molecular Formula C34H26N6Na2O6S2
Molecular Weight (g/mol) 724.718
InChI Key SUXCALIDMIIJCK-UHFFFAOYSA-L
Synonym benzopurpurine 4b, benzopurpurin 4b, direct red 2, c.i. direct red 2, amanil purpurine, diphenyl red 4b, hispamin red 4b, direct red dcb, azocard red 4b, diazine red 4b
PubChem CID 13816
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]-3-methylphenyl]-2-methylphenyl]diazenyl]naphthalene-1-sulfonate
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N=NC3=C(C4=CC=CC=C4C(=C3)S(=O)(=O)[O-])N)C)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N.[Na+].[Na+]

Specifications

Quantity 25 g
Formula Weight 724.71
Chemical Name or Material Benzopurpurin B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.