CAS RN 99210-90-9
CAS RN 99210-90-9
IUPAC Name:
2-methylpentane-2,4-diol
Synonyms:
2-methyl-2,4-pentanediol hexylene glycol, titanium(4+) salt caswell no. 574 titanium(4+) salt hexylene glycol epa pesticide chemical code 068601 MPD hexylene glycol (substance) hexylene glycol pinakon diolane
Molecular Weight (g/mol):
118.18
Molecular Formula:
C6H14O2
InChi Key:
SVTBMSDMJJWYQN-UHFFFAOYNA-N
SMILES:
CC(O)CC(C)(C)O
1
–
1
of
1
results
1
(R)-(-)-2-Methyl-2,4-pentanediol, 98+%
CAS: 99210-90-9 Molecular Formula: C6H14O2 Molecular Weight (g/mol): 118.18 MDL Number: MFCD00064275 InChI Key: SVTBMSDMJJWYQN-UHFFFAOYNA-N Synonym: 4r-2-methylpentane-2,4-diol,r---2-methyl-2,4-pentanediol,mrd,2-methylpentan-2,4-diol,r-2-methyl-2,4-pentanediol,1hty,r-mpd,4-methyl-2,4-pentanediole,r---2-methyl-2,4-pentandiol,.+/-.-2-methyl-2,4-pentanediol PubChem CID: 5288845 IUPAC Name: (4R)-2-methylpentane-2,4-diol SMILES: CC(O)CC(C)(C)O