Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2-Methyl-2,4-pentanediol, 98+%

Catalog No. AAL1452303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1452303 1 g
AAL14523MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1452303 Supplier Thermo Scientific Chemicals Supplier No. L1452303
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 99210-90-9 | C6H14O2 | 118.18 g/mol

(R)-(-)-2-Methyl-2,4-pentanediol is used as a chiral reagent. Also used as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-2-Methyl-2,4-pentanediol is used as a chiral reagent. Also used as intermediate for pharmaceutical.

Solubility
Fully miscible with water.

Notes
Hygroscopic, store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99210-90-9
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00064275
InChI Key SVTBMSDMJJWYQN-UHFFFAOYNA-N
Synonym 4r-2-methylpentane-2,4-diol, r---2-methyl-2,4-pentanediol, mrd, 2-methylpentan-2,4-diol, r-2-methyl-2,4-pentanediol, 1hty, r-mpd, 4-methyl-2,4-pentanediole, r---2-methyl-2,4-pentandiol, .+/-.-2-methyl-2,4-pentanediol
PubChem CID 5288845
IUPAC Name (4R)-2-methylpentane-2,4-diol
SMILES CC(O)CC(C)(C)O

Specifications

Density 0.92
Boiling Point 197°C
Flash Point 93°C (199°F)
Refractive Index 1.4275
Quantity 1 g
Beilstein 6051650
Sensitivity Hygroscopic
Solubility Information Fully miscible with water.
Optical Rotation −21° (Neat)
Formula Weight 118.18
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-2-Methyl-2,4-pentanediol
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.