accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 993-22-6

IUPAC Name: azide; tetrabutylazanium
Synonyms: azide; tetrabutylammonium ion
Molecular Weight (g/mol): 284.49
Molecular Formula: C16H36N4
InChi Key: GMRIOAVKKGNMMV-UHFFFAOYSA-N
SMILES: [N-]=[N+]=[N-].CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results