Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Azide 95.0+%, TCI America™
SDP

Catalog No. T09205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09205G 5 g
T092025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09205G Supplier TCI America Supplier No. T09205G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 993-22-6
Molecular Formula C16H36N4
Molecular Weight (g/mol) 284.492
MDL Number MFCD00060069
InChI Key GMRIOAVKKGNMMV-UHFFFAOYSA-N
PubChem CID 5743232
IUPAC Name tetrabutylazanium;azide
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[N-]=[N+]=[N-]

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 284.49
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Azide
EINECSNumber (2)-0186
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.