accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNNNOOOH

CAS RN 99310-01-7

IUPAC Name: 3-{1-[(benzyloxy)methyl]-1H-imidazol-5-yl}-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: 3-{3-[(benzyloxy)methyl]imidazol-4-yl}-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 375.43
Molecular Formula: C19H25N3O5
InChi Key: LPVKZCHCZSFTOJ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CN=CN1COCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)