accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99365-69-2

IUPAC Name: 7-nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Synonyms: 7-nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Molecular Weight (g/mol): 214.65
Molecular Formula: C9H11ClN2O2
InChi Key: KGIXLJRTYZOUCW-UHFFFAOYSA-N
SMILES: Cl.[O-][N+](=O)C1=CC2=C(CCNC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results