accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COH3C

CAS RN 994-05-8

IUPAC Name: 2-methoxy-2-methylbutane
Synonyms: tert-amyl methyl ether methyl-T-amyl ether
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: HVZJRWJGKQPSFL-UHFFFAOYSA-N
SMILES: CCC(C)(C)OC
Molecular Weight (g/mol) 
  • (3)
  • (1)
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)