Learn More
Description
- Tert-amyl methyl ether is an fuel ether oxygenate and has been quantitated in water by membrane extraction ion mobility spectrometry
- Tert-amyl methyl ether is also referred to as 2-methoxy-2-methyl butane
- Kinetics of ion exchange resin (amberlyst 18) catalyzed tert-amyl methyl ether reactions have been investigated
Specifications
Specifications
| CAS | 994-05-8 |
| Density | 0.76-0.78 g/mL (at 25°C (literature)) |
| Boiling Point | 85°C to 86°C (lit.) |
| Molecular Formula | C6H14O |
| Linear Formula | C2H5C(CH3)2 OCH3 |
| Refractive Index | n20/D 1.389 (literature); n20/D 1.3896 |
| Quantity | 5 mL |
| MDL Number | MFCD00010186 |
| Synonym | 1,1-Dimethylpropyl methyl ether; Methyl 2-methyl-2-butyl ether; Methyl tert-pentyl ether; TAME |
| Molecular Weight (g/mol) | 102.17 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.