accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiClH3CH3C

CAS RN 994-30-9

IUPAC Name: chlorotriethylsilane
Synonyms: chlorotriethylsilane
Molecular Weight (g/mol): 150.72
Molecular Formula: C6H15ClSi
InChi Key: DCFKHNIGBAHNSS-UHFFFAOYSA-N
SMILES: CC[Si](Cl)(CC)CC
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (9)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)