Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorotriethylsilane

Catalog No. 2350675G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-506-75G 5 g
23-506-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 23-506-75G Supplier Sigma Aldrich Supplier No. 2350675G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Employed together with Grignard reagents in a palladium-catalyzed carbosilylation of allenes. The Grignard attacks the allene central carbon and the terminal carbon is silylated.

Specifications

CAS 994-30-9
Density 0.898 g/mL at 25°C
Boiling Point 142°C to 144°C
Molecular Formula C6H15ClSi
Refractive Index n20/D 1.43
Linear Formula (C2H5)3SiCl
MDL Number MFCD00000507
Quantity 5 g
Synonym TESCl; Triethylchlorosilane
Molecular Weight (g/mol) 150.72 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.