accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 994-49-0

IUPAC Name: triethyl[(triethylsilyl)oxy]silane
Synonyms: disiloxane, hexaethyl-
Molecular Weight (g/mol): 246.54
Molecular Formula: C12H30OSi2
InChi Key: WILBTFWIBAOWLN-UHFFFAOYSA-N
SMILES: CC[Si](CC)(CC)O[Si](CC)(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results