Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexaethyldisiloxane, 99%

Catalog No. AAL0436706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0436706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0436706 Supplier Thermo Scientific Chemicals Supplier No. L0436706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 994-49-0 | C12H30OSi2 | 246.541 g/mol

Hexaethyldisiloxane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaethyldisiloxane is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well seale
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 994-49-0
Molecular Formula C12H30OSi2
Molecular Weight (g/mol) 246.541
MDL Number MFCD00026693
InChI Key WILBTFWIBAOWLN-UHFFFAOYSA-N
Synonym hexaethyldisiloxane, disiloxane, hexaethyl, 1,1,1,3,3,3-hexaethyldisiloxane, hexaethyl disiloxane, disiloxane, 1,1,1,3,3,3-hexaethyl, triethylsilylether, triethylsilyl ether, hexaethyldisiloxane min, triethyl triethylsilyloxy silane, 1,1,3,3,3-hexaethyldisiloxane
PubChem CID 70442
IUPAC Name triethyl(triethylsilyloxy)silane
SMILES CC[Si](CC)(CC)O[Si](CC)(CC)CC

Specifications

Boiling Point 231°C
Melting Point -115°C
Quantity 5 g
Flash Point 87°C (188°F)
Refractive Index 1.433
Density 0.84
Linear Formula (CH3CH2)3SiOSi(CH2CH3)3
Beilstein 1750865
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 246.54
Percent Purity 99%
Chemical Name or Material Hexaethyldisiloxane
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 213-619-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.