accessibility menu, dialog, popup

UserName

Viewing profile as user:
OClONH2H2NHClHCl

CAS RN 99503-73-8

IUPAC Name: 3-[3-(3-aminophenoxy)-5-chlorophenoxy]aniline dihydrochloride
Synonyms: 3-[3-(3-aminophenoxy)-5-chlorophenoxy]aniline dihydrochloride
Molecular Weight (g/mol): 399.7
Molecular Formula: C18H17Cl3N2O2
InChi Key: YZUFDVOYHZOIGU-UHFFFAOYSA-N
SMILES: Cl.Cl.NC1=CC(OC2=CC(OC3=CC=CC(N)=C3)=CC(Cl)=C2)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results